Assay Details
| Gene Symbol: Gckr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144909.1 | NP_659158.1 | 16 - 17 | 1460 | 90 | |
| GenBank mRNA | BC012412.1 | - | 16 - 17 | 1460 | 4159892 | 90 |
More Information
Updated Mapping Information
Set Membership: |
|

Gckr
Genomic Map