Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500024556.mm.1 |
TC0500002790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024556.mm.1 |
NM_001285511.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024556.mm.1 |
NM_001285514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024556.mm.1 |
NM_144910.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024556.mm.1 |
NM_178854.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024557.mm.1 |
TC0500002790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500024557.mm.1 |
NM_001285511.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024557.mm.1 |
NM_001285514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024557.mm.1 |
NM_144910.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500024557.mm.1 |
NM_178854.4 |
Refseq |