Assay Details
| Gene Symbol: Rad9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144912.3 | NP_659161.2 | 2 - 3 | 127 | 78 | |
| GenBank mRNA | AY297460.1 | - | 2 - 3 | 127 | 78 | |
| BC021784.1 | - | 2 - 3 | 127 | 4162418 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rad9b
Genomic Map