Assay Details
| Gene Symbol: Rad9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144912.3 | NP_659161.2 | 6 - 7 | 595 | 80 | |
| GenBank mRNA | AK136504.1 | - | 6 - 7 | 631 | 80 | |
| AY297460.1 | - | 6 - 7 | 595 | 80 | ||
| BC021784.1 | - | 6 - 7 | 595 | 4162418 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049900 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049901 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513013 | AY297460.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513013 | BC021784.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513013 | AK136504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513013 | NM_144912.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513013 | AY297460.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513013 | BC021784.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513013 | AK136504.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513013 | NM_144912.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215667 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 117189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514787 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935171 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 567652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 617136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154450 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83995 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 125594 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74507 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145827 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11032 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152661 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116298 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513013 | AY297460.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513013 | BC021784.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513013 | AK136504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513013 | NM_144912.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513013 | AY297460.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513013 | BC021784.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513013 | AK136504.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513013 | NM_144912.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049870 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24049899 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05724285 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01207260 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152661 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01207269 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116298 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01207278 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01207288 | - | - |
| Whole Mouse Genome G4122A | A_51_P513013 | AY297460.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513013 | BC021784.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513013 | AK136504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513013 | NM_144912.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513013 | AY297460.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513013 | BC021784.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513013 | AK136504.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P513013 | NM_144912.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106341 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152661 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116298 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01207278 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04782185 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04430154 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04782182 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04430155 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04782194 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049901 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049900 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24049898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24049888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04686895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04686909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04750120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049896 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04779435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049870 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04686875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04779439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049748 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04686878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049899 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04779445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04686900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049884 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24049874 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10533468 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533464 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533471 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533466 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533470 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533467 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533473 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533472 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533469 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533465 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10533463 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425800_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439955_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10533462 | - | - |
| Mouse Genome 430 2.0 Array | 1439955_at | - | - |
| Mouse Genome 430 2.0 Array | 1439955_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425800_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425800_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231724_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231724_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500028657.mm.1 | TC0500003206.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500028657.mm.1 | NM_144912.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500028657.mm.1 | NM_144912.3 | Refseq |
| Mouse Expression Set 430 | 1425800_at | - | - |
| Mouse Expression Set 430 | 1425800_PM_at | - | - |
| Mouse Expression Set 430 | 1439955_at | - | - |
| Mouse Expression Set 430 | 1439955_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425800_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425800_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425800_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425800_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439955_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1439955_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 558044 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2320064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002320064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225080 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225080 | - | - |
| mouse-6 v1.1 (Illumina) | scl0231724.1_307-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2320064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0231724.1_307-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2320064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1225080 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1225080 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0231724.1_307-S | - | - |

Rad9b
Genomic Map