Assay Details
| Gene Symbol: Mepce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144913.3 | NP_659162.3 | 2 - 3 | 2464 | 72 | |
| GenBank mRNA | AK139731.1 | - | 2 - 3 | 696 | 72 | |
| AK154415.1 | - | 2 - 3 | 2427 | 72 | ||
| BC017157.1 | - | 2 - 3 | 733 | 72 | ||
| BC026876.1 | - | 2 - 3 | 1933 | 3662118 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mepce
Genomic Map