Assay Details
| Gene Symbol: Daglb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144915.3 | NP_659164.2 | 9 - 10 | 1410 | 61 | |
| GenBank mRNA | AK081509.1 | - | 48 | 61 | ||
| AK086747.1 | - | 9 - 10 | 1322 | 61 | ||
| AK087884.1 | - | 9 - 10 | 1265 | 61 | ||
| AK136372.1 | - | 9 - 10 | 1268 | 61 | ||
| BC016105.1 | - | 9 - 10 | 1306 | 4921222 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Daglb
Genomic Map