Assay Details
| Gene Symbol: Atp1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290469.1 | NP_001277398.1 | 21 - 22 | 3104 | 104 | |
| GenBank mRNA | BC020177.1 | - | 21 - 22 | 3062 | 4506376 | 104 |
| BC027000.1 | - | 14 - 15 | 2124 | 104 | ||
| BC027114.1 | - | 9 - 10 | 1233 | 104 | ||
| BC034645.1 | - | 21 - 22 | 3060 | 5357731 | 104 | |
| BC037206.1 | - | 21 - 22 | 3059 | 5362423 | 104 | |
| BC040512.1 | - | 6 - 7 | 737 | 104 | ||
| BC042894.1 | - | 21 - 22 | 3104 | 5364812 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atp1a3
Genomic Map