Assay Details
| Gene Symbol: Ces1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144930.2 | NP_659179.1 | 6 - 7 | 830 | 85 | |
| GenBank mRNA | AB023632.1 | - | 2 - 3 | 233 | 85 | |
| AI786051.1 | - | 2 - 3 | 226 | 85 | ||
| AK136491.1 | - | 6 - 7 | 822 | 85 | ||
| BC013479.1 | - | 6 - 7 | 805 | 4239756 | 85 | |
| EH107325.1 | - | 1 - 2 | 78 | 85 | ||
More Information
Set Membership: |
|

Ces1f
Genomic Map