Assay Details
| Gene Symbol: Ces1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144930.2 | NP_659179.1 | 10 - 11 | 1196 | 92 | |
| GenBank mRNA | AB023632.1 | - | 6 - 7 | 599 | 92 | |
| AK136491.1 | - | 10 - 11 | 1188 | 92 | ||
| BC013479.1 | - | 10 - 11 | 1171 | 4239756 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ces1f
Genomic Map