Assay Details
| Gene Symbol: Prc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285997.1 | NP_001272926.1 | 7 - 8 | 1098 | 83 | |
| NM_001285998.1 | NP_001272927.1 | 7 - 8 | 1098 | 83 | ||
| NM_145150.3 | NP_660132.2 | 7 - 8 | 1098 | 83 | ||
| GenBank mRNA | AK029140.1 | - | 7 - 8 | 1085 | 83 | |
| AK088576.1 | - | 7 - 8 | 1070 | 83 | ||
| AK135778.1 | - | 5 - 6 | 723 | 83 | ||
| BC005475.1 | - | 7 - 8 | 1032 | 3592138 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prc1
Genomic Map