Assay Details
| Gene Symbol: Wasf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145155.3 | NP_660137.1 | 3 - 4 | 560 | 117 | |
| GenBank mRNA | AF454703.1 | - | 2 - 3 | 179 | 117 | |
| AK044481.1 | - | 581 | 117 | |||
| BC027038.1 | - | 3 - 4 | 560 | 5363785 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wasf3
Genomic Map