Assay Details
| Gene Symbol: Wasf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145155.3 | NP_660137.1 | 8 - 9 | 1410 | 126 | |
| GenBank mRNA | AF454703.1 | - | 7 - 8 | 1029 | 126 | |
| AK044481.1 | - | 1431 | 126 | |||
| BC027038.1 | - | 8 - 9 | 1410 | 5363785 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wasf3
Genomic Map