Assay Details
| Gene Symbol: Dars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177445.5 | NP_803228.2 | 7 - 8 | 755 | 97 | |
| GenBank mRNA | AK077533.1 | - | 7 - 8 | 678 | 97 | |
| AK144470.1 | - | 7 - 8 | 700 | 97 | ||
| AK146350.1 | - | 7 - 8 | 699 | 97 | ||
| AK151027.1 | - | 7 - 8 | 699 | 97 | ||
| AK151897.1 | - | 7 - 8 | 666 | 97 | ||
| AK152304.1 | - | 7 - 8 | 666 | 97 | ||
| AK152562.1 | - | 7 - 8 | 666 | 97 | ||
| AK153256.1 | - | 7 - 8 | 666 | 97 | ||
| AK153299.1 | - | 7 - 8 | 666 | 97 | ||
| AK166009.1 | - | 7 - 8 | 666 | 97 | ||
| AK169247.1 | - | 7 - 8 | 666 | 97 | ||
| AK169716.1 | - | 7 - 8 | 643 | 97 | ||
| AK193968.1 | - | 3 - 4 | 212 | 97 | ||
| BC008638.1 | - | 7 - 8 | 653 | 3586278 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dars
Genomic Map