Assay Details
| Gene Symbol: Dars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177445.5 | NP_803228.2 | 9 - 10 | 1005 | 113 | |
| GenBank mRNA | AK077533.1 | - | 9 - 10 | 928 | 113 | |
| AK146350.1 | - | 9 - 10 | 949 | 113 | ||
| AK151027.1 | - | 9 - 10 | 949 | 113 | ||
| AK151897.1 | - | 9 - 10 | 916 | 113 | ||
| AK152304.1 | - | 9 - 10 | 916 | 113 | ||
| AK152562.1 | - | 9 - 10 | 916 | 113 | ||
| AK153256.1 | - | 9 - 10 | 916 | 113 | ||
| AK153299.1 | - | 9 - 10 | 916 | 113 | ||
| AK169247.1 | - | 9 - 10 | 916 | 113 | ||
| AK169716.1 | - | 9 - 10 | 893 | 113 | ||
| BC008638.1 | - | 9 - 10 | 903 | 3586278 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dars
Genomic Map