Assay Details
| Gene Symbol: Wdr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145514.5 | NP_663489.4 | 5 - 6 | 1339 | 90 | |
| GenBank mRNA | AK075743.1 | - | 5 - 6 | 731 | 90 | |
| AK082326.1 | - | 6 - 7 | 634 | 90 | ||
| BC020044.1 | - | 1 - 2 | 84 | 90 | ||
| BC058601.1 | - | 3 - 4 | 335 | 90 | ||
| BC094233.1 | - | 2 - 3 | 224 | 90 | ||
| BC139336.1 | - | 5 - 6 | 501 | 8862358 | 90 | |
| BC139337.1 | - | 5 - 6 | 501 | 8862359 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr26
Genomic Map