Assay Details
| Gene Symbol: Ndufs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160038.1 | NP_001153510.1 | 4 - 5 | 653 | 69 | |
| NM_001160039.1 | NP_001153511.1 | 4 - 5 | 495 | 69 | ||
| NM_001160040.1 | NP_001153512.1 | 3 - 4 | 549 | 69 | ||
| NM_145518.2 | NP_663493.2 | 3 - 4 | 391 | 69 | ||
| GenBank mRNA | AK034597.1 | - | 3 - 4 | 460 | 69 | |
| AK036926.1 | - | 3 - 4 | 256 | 69 | ||
| AK078590.1 | - | 3 - 4 | 237 | 69 | ||
| AK142711.1 | - | 3 - 4 | 271 | 69 | ||
| AK146814.1 | - | 3 - 4 | 409 | 69 | ||
| AK166604.1 | - | 4 - 5 | 342 | 69 | ||
| AK167705.1 | - | 3 - 4 | 239 | 69 | ||
| BC006660.1 | - | 3 - 4 | 278 | 3501043 | 69 | |
| BC015300.1 | - | 3 - 4 | 211 | 4236882 | 69 | |
| BG808119.1 | - | 3 - 4 | 389 | 69 | ||
| BU708393.1 | - | 4 - 5 | 496 | 69 | ||
| BY709888.1 | - | 3 - 4 | 392 | 69 | ||
| CN715066.1 | - | 3 - 4 | 267 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndufs1
Genomic Map