Assay Details
| Gene Symbol: Rabepk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145522.4 | NP_663497.2 | 6 - 7 | 847 | 108 | |
| GenBank mRNA | AK035343.1 | - | 6 - 7 | 727 | 108 | |
| BC019800.1 | - | 6 - 7 | 723 | 5150243 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rabepk
Genomic Map