Assay Details
| Gene Symbol: Mall | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145532.3 | NP_663507.1 | 2 - 3 | 306 | 81 | |
| GenBank mRNA | AK078971.1 | - | 2 - 3 | 307 | 81 | |
| AK163867.1 | - | 2 - 3 | 307 | 81 | ||
| BC012256.1 | - | 2 - 3 | 341 | 4205488 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mall
Genomic Map