Assay Details
| Gene Symbol: Rap1gds1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040690.2 | NP_001035780.1 | 10 - 11 | 1336 | 67 | |
| NM_001286759.1 | NP_001273688.1 | 10 - 11 | 1336 | 67 | ||
| NM_145544.3 | NP_663519.2 | 9 - 10 | 1189 | 67 | ||
| GenBank mRNA | AK052125.1 | - | 9 - 10 | 1137 | 67 | |
| AK154852.1 | - | 10 - 11 | 1322 | 67 | ||
| AK155793.1 | - | 10 - 11 | 1322 | 67 | ||
| AK160989.1 | - | 10 - 11 | 1272 | 67 | ||
| AK166315.1 | - | 10 - 11 | 1269 | 67 | ||
| AK172052.1 | - | 10 - 11 | 1338 | 67 | ||
| BC011279.1 | - | 9 - 10 | 1058 | 4008751 | 67 | |
| CN702379.1 | - | 1 - 2 | 149 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rap1gds1
Genomic Map