Assay Details
Gene Symbol: Cyp2j13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145548.4 | NP_663523.2 | 1 - 2 | 275 | 106 | |
GenBank mRNA | AK143903.1 | - | 1 - 2 | 276 | 106 | |
AK143981.1 | - | 1 - 2 | 272 | 106 | ||
AK162358.1 | - | 279 | 106 | |||
AK165405.1 | - | 1 - 2 | 275 | 106 | ||
AK165469.1 | - | 1 - 2 | 253 | 106 | ||
AK165473.1 | - | 1 - 2 | 253 | 106 | ||
BC016446.1 | - | 1 - 2 | 265 | 4235131 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
