Assay Details
Gene Symbol: Cyp2j13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145548.4 | NP_663523.2 | 3 - 4 | 591 | 144 | |
GenBank mRNA | AK143903.1 | - | 3 - 4 | 592 | 144 | |
AK143981.1 | - | 3 - 4 | 588 | 144 | ||
AK162358.1 | - | 595 | 144 | |||
AK165405.1 | - | 3 - 4 | 591 | 144 | ||
AK165469.1 | - | 3 - 4 | 569 | 144 | ||
AK165473.1 | - | 3 - 4 | 569 | 144 | ||
BC016446.1 | - | 3 - 4 | 581 | 4235131 | 144 | |
BC024451.1 | - | 2 - 3 | 211 | 144 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC024451.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
