Assay Details
| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145551.4 | NP_663526.3 | 8 - 9 | 1026 | 62 | |
| GenBank mRNA | AK029158.1 | - | 8 - 9 | 1027 | 62 | |
| AK033425.1 | - | 7 - 8 | 956 | 62 | ||
| AK050696.1 | - | 7 - 8 | 906 | 62 | ||
| BC021357.1 | - | 8 - 9 | 1007 | 5012356 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a9
Genomic Map