Assay Details
| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145551.4 | NP_663526.3 | 9 - 10 | 1160 | 100 | |
| GenBank mRNA | AK029158.1 | - | 9 - 10 | 1161 | 100 | |
| AK033425.1 | - | 8 - 9 | 1090 | 100 | ||
| AK050696.1 | - | 8 - 9 | 1040 | 100 | ||
| BC021357.1 | - | 9 - 10 | 1141 | 5012356 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a9
Genomic Map