Assay Details
| Gene Symbol: Slc5a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145551.4 | NP_663526.3 | 12 - 13 | 1589 | 72 | |
| GenBank mRNA | AK029158.1 | - | 12 - 13 | 1590 | 72 | |
| AK033425.1 | - | 11 - 12 | 1519 | 72 | ||
| AK050696.1 | - | 11 - 12 | 1469 | 72 | ||
| BC021357.1 | - | 12 - 13 | 1570 | 5012356 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a9
Genomic Map