Assay Details
| Gene Symbol: Siglecf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271019.1 | NP_001257948.1 | 5 - 6 | 831 | 99 | |
| NM_145581.2 | NP_663556.1 | 5 - 6 | 831 | 99 | ||
| GenBank mRNA | AF293371.1 | - | 5 - 6 | 767 | 99 | |
| AK087658.1 | - | 4 - 5 | 894 | 99 | ||
| AK146203.1 | - | 4 - 5 | 868 | 99 | ||
| AK155469.1 | - | 5 - 6 | 778 | 99 | ||
| AY210401.1 | - | 4 - 5 | 727 | 99 | ||
| BC145038.1 | - | 4 - 5 | 762 | 9053586 | 99 | |
| BC145987.1 | - | 4 - 5 | 762 | 40131290 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Siglecf
Genomic Map