Assay Details
Gene Symbol: Siglecf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271019.1 | NP_001257948.1 | 8 - 9 | 1522 | 90 | |
NM_145581.2 | NP_663556.1 | 9 - 10 | 1624 | 90 | ||
GenBank mRNA | AF293371.1 | - | 9 - 10 | 1560 | 90 | |
AK146203.1 | - | 8 - 9 | 1661 | 90 | ||
AK155469.1 | - | 9 - 10 | 1573 | 90 | ||
BC145038.1 | - | 7 - 8 | 1453 | 9053586 | 90 | |
BC145987.1 | - | 8 - 9 | 1555 | 40131290 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043277 | AK155469.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043277 | AK146203.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043277 | NM_145581.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043277 | AF293371.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115406 | BC145987.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115406 | AK155469.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115406 | AY210401.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115406 | BC145038.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115406 | AK146203.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105064 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494077 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616040 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376535 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 1299 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130511 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115406 | BC145987.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115406 | AK155469.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115406 | AY210401.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115406 | BC145038.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115406 | AK146203.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043277 | AK155469.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043277 | AK146203.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043277 | NM_145581.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043277 | AF293371.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P199475 | BC145987.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P199475 | AK155469.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P199475 | BC145038.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P199475 | AK087658.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P199475 | AK146203.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081935 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081929 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081951 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081927 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081939 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081943 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081937 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081945 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081950 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081953 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081940 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081941 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081948 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081933 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081955 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081959 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081961 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081949 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081930 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25081925 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130511 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01504623 | - | - |
Whole Mouse Genome G4122A | A_51_P199475 | BC145987.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P199475 | AK155469.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P199475 | BC145038.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P199475 | AK087658.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P199475 | AK146203.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103181 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130511 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06431963 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06444852 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06431967 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06431970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081969 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081977 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081961 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081966 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081970 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081939 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25082004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25081968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081988 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081975 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05983204 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06431961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06052095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06052080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06043739 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081966 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081941 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06431969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081968 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081985 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081908 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25082006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081929 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081951 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05930193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06469954 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081924 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081983 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081915 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05895796 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081992 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05895787 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081919 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081981 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081953 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081939 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081958 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081910 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06444852 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06431954 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081987 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081979 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081984 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081971 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06474220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081973 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081926 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05895789 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06052075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25081922 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06052087 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10552376 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552375 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552374 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552373 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552379 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552370 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552378 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552371 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552377 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10552372 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1424975_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10552369 | - | - |
Mouse Genome 430 2.0 Array | 1424975_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1424975_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233186_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 233186_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700006090.mm.1 | TC0700000756.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700006090.mm.1 | NM_001271019.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006090.mm.1 | NM_145581.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006092.mm.1 | TC0700000756.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700006092.mm.1 | NM_001271019.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700006092.mm.1 | NM_145581.2 | Refseq |
Mouse Expression Set 430 | 1424975_PM_at | - | - |
Mouse Expression Set 430 | 1424975_at | - | - |
Murine Genome U74v2 Set | 166745_i_at | - | - |
Murine Genome U74v2 Set | 166745_PM_i_at | - | - |
Mouse Genome 430A 2.0 Array | 1424975_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1424975_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424975_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1424975_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 356847 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 5690398 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005690398 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2675309 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2675309 | - | - |
mouse-6 v1.1 (Illumina) | scl32759.9.1_89-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5690398 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl32759.9.1_89-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5690398 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2675309 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2675309 | - | - |
