Assay Details
| Gene Symbol: BC021891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145608.2 | NP_663583.2 | 3 - 4 | 1318 | 61 | |
| GenBank mRNA | AK220424.1 | - | 3 - 4 | 1318 | 61 | |
| BC021891.1 | - | 3 - 4 | 1314 | 3156324 | 61 | |
More Information
Set Membership: |
|

BC021891
Genomic Map