Assay Details
| Gene Symbol: Lin28a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145833.1 | NP_665832.1 | 2 - 3 | 306 | 75 | |
| GenBank mRNA | AB221669.1 | - | 2 - 3 | 231 | 75 | |
| AF521097.1 | - | 2 - 3 | 306 | 75 | ||
| BC068304.1 | - | 2 - 3 | 263 | 30432006 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lin28a
Genomic Map