| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400030589.mm.1 |
TC0400003723.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030589.mm.1 |
NM_145833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030589.mm.1 |
NM_145833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030591.mm.1 |
TC0400003723.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030591.mm.1 |
NM_145833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030591.mm.1 |
NM_145833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030592.mm.1 |
TC0400003723.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400030592.mm.1 |
NM_145833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400030592.mm.1 |
NM_145833.1 |
Refseq |