Assay Details
| Gene Symbol: Hmgcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145942.5 | NP_666054.2 | 2 - 3 | 222 | 62 | |
| GenBank mRNA | AK031297.1 | - | 2 - 3 | 111 | 62 | |
| AK044835.1 | - | 2 - 3 | 112 | 62 | ||
| AK136171.1 | - | 2 - 3 | 111 | 62 | ||
| BC023851.1 | - | 2 - 3 | 138 | 5347038 | 62 | |
| BC031363.1 | - | 1 - 2 | 70 | 5357048 | 62 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK031297.1,AK044835.1,AK136171.1,BC023851.1,BC031363.1,NM_145942.5 transcripts.
Set Membership: |
|

Hmgcs1
Genomic Map