Assay Details
| Gene Symbol: Hmgcs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291439.1 | NP_001278368.1 | 7 - 8 | 1357 | 111 | |
| NM_145942.5 | NP_666054.2 | 8 - 9 | 1416 | 111 | ||
| GenBank mRNA | AK031297.1 | - | 8 - 9 | 1305 | 111 | |
| AK044835.1 | - | 8 - 9 | 1306 | 111 | ||
| AK045094.1 | - | 7 - 8 | 1245 | 111 | ||
| AK135551.1 | - | 7 - 8 | 1245 | 111 | ||
| AK136171.1 | - | 8 - 9 | 1305 | 111 | ||
| AK144384.1 | - | 7 - 8 | 1247 | 111 | ||
| AK146042.1 | - | 7 - 8 | 1247 | 111 | ||
| AK146590.1 | - | 7 - 8 | 1247 | 111 | ||
| AK191228.1 | - | 2 - 3 | 276 | 111 | ||
| AK201868.1 | - | 2 - 3 | 275 | 111 | ||
| BC023851.1 | - | 8 - 9 | 1332 | 5347038 | 111 | |
| BC029693.1 | - | 7 - 8 | 1262 | 5375374 | 111 | |
| BC031363.1 | - | 7 - 8 | 1264 | 5357048 | 111 | |
| BC034317.1 | - | 6 - 7 | 1208 | 5366786 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmgcs1
Genomic Map