Assay Details
| Gene Symbol: Slc26a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145947.2 | NP_666059.2 | 1 - 2 | 92 | 100 | |
| GenBank mRNA | AK054327.1 | - | 1 - 2 | 93 | 100 | |
| BC026928.1 | - | 1 - 2 | 71 | 4020760 | 100 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC026928.1,NM_145947.2 transcripts.
Set Membership: |
|

Slc26a7
Genomic Map