Assay Details
| Gene Symbol: Slc26a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145947.2 | NP_666059.2 | 5 - 6 | 766 | 124 | |
| GenBank mRNA | AF345194.1 | - | 4 - 5 | 668 | 124 | |
| AK054327.1 | - | 5 - 6 | 767 | 124 | ||
| BC026928.1 | - | 5 - 6 | 745 | 4020760 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a7
Genomic Map