Assay Details
| Gene Symbol: Slc26a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145947.2 | NP_666059.2 | 8 - 9 | 1154 | 90 | |
| GenBank mRNA | AF345194.1 | - | 7 - 8 | 1056 | 90 | |
| AK054327.1 | - | 8 - 9 | 1155 | 90 | ||
| BC026928.1 | - | 8 - 9 | 1133 | 4020760 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a7
Genomic Map