Assay Details
| Gene Symbol: Cc2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145970.1 | NP_666082.2 | 12 - 13 | 1380 | 83 | |
| GenBank mRNA | BC016188.1 | - | 12 - 13 | 1380 | 4482793 | 83 |
| BC027028.1 | - | 11 - 12 | 1195 | 5363547 | 83 | |
More Information
Set Membership: |
|

Cc2d1a
Genomic Map