Assay Details
| Gene Symbol: Cc2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145970.1 | NP_666082.2 | 19 - 20 | 2099 | 65 | |
| GenBank mRNA | BC016188.1 | - | 19 - 20 | 2099 | 4482793 | 65 |
| BC027028.1 | - | 18 - 19 | 1914 | 5363547 | 65 | |
| BC030439.1 | - | 3 - 4 | 206 | 65 | ||
| DQ329239.1 | - | 12 - 13 | 1149 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cc2d1a
Genomic Map