Assay Details
| Gene Symbol: Ddx46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282055.1 | NP_001268984.1 | 14 - 15 | 1967 | 83 | |
| GenBank mRNA | AK088950.1 | - | 13 - 14 | 3557 | 83 | |
| AK129220.1 | - | 13 - 14 | 3558 | 83 | ||
| AK134073.1 | - | 2 - 3 | 1213 | 83 | ||
| BC026492.1 | - | 7 - 8 | 912 | 4505095 | 83 | |
| BC092240.1 | - | 13 - 14 | 1878 | 30662573 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx46
Genomic Map