Assay Details
| Gene Symbol: Ddx46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282055.1 | NP_001268984.1 | 21 - 22 | 3158 | 75 | |
| GenBank mRNA | AK129220.1 | - | 20 - 21 | 4749 | 75 | |
| AK160322.1 | - | 4 - 5 | 631 | 75 | ||
| BC026492.1 | - | 14 - 15 | 2103 | 4505095 | 75 | |
| BC092240.1 | - | 20 - 21 | 3069 | 30662573 | 75 | |
More Information
Set Membership: |
|

Ddx46
Genomic Map