Assay Details
| Gene Symbol: L3mbtl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289711.1 | NP_001276640.1 | 7 - 8 | 967 | 70 | |
| NM_001289712.1 | NP_001276641.1 | 7 - 8 | 967 | 70 | ||
| NM_145993.5 | NP_666105.2 | 7 - 8 | 967 | 70 | ||
| NR_110362.1 | 7 - 8 | 967 | 70 | |||
| GenBank mRNA | AK029115.1 | - | 7 - 8 | 912 | 70 | |
| AK036510.1 | - | 7 - 8 | 920 | 70 | ||
| AK132012.1 | - | 7 - 8 | 920 | 70 | ||
| AK141810.1 | - | 6 - 7 | 2101 | 70 | ||
| AK145560.1 | - | 7 - 8 | 909 | 70 | ||
| AK146632.1 | - | 7 - 8 | 923 | 70 | ||
| AK156236.1 | - | 7 - 8 | 922 | 70 | ||
| AY237000.1 | - | 7 - 8 | 900 | 70 | ||
| AY237001.1 | - | 7 - 8 | 900 | 70 | ||
| AY237002.1 | - | 7 - 8 | 900 | 70 | ||
| BC023933.1 | - | 7 - 8 | 908 | 5325131 | 70 | |
| BC030864.1 | - | 7 - 8 | 870 | 4211489 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

L3mbtl2
Genomic Map