Assay Details
| Gene Symbol: Rhot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145999.2 | NP_666111.1 | 8 - 9 | 956 | 108 | |
| GenBank mRNA | AY311390.1 | - | 8 - 9 | 956 | 108 | |
| BC029777.1 | - | 8 - 9 | 589 | 4950061 | 108 | |
| BC034062.1 | - | 8 - 9 | 611 | 4207654 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhot2
Genomic Map