Assay Details
| Gene Symbol: Rhot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145999.2 | NP_666111.1 | 11 - 12 | 1291 | 72 | |
| GenBank mRNA | AK142183.1 | - | 8 - 9 | 1198 | 72 | |
| AY311390.1 | - | 11 - 12 | 1291 | 72 | ||
| BC023163.1 | - | 1 - 2 | 61 | 72 | ||
| BC029777.1 | - | 11 - 12 | 924 | 4950061 | 72 | |
| BC034062.1 | - | 11 - 12 | 946 | 4207654 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhot2
Genomic Map