Assay Details
Gene Symbol: Rhot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145999.2 | NP_666111.1 | 16 - 17 | 1830 | 58 | |
GenBank mRNA | AK216218.1 | - | 2 - 3 | 174 | 58 | |
AY311390.1 | - | 16 - 17 | 1830 | 58 | ||
BC023163.1 | - | 6 - 7 | 600 | 58 | ||
BC029777.1 | - | 16 - 17 | 1463 | 4950061 | 58 | |
BC034062.1 | - | 16 - 17 | 1485 | 4207654 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
