Assay Details
| Gene Symbol: Hip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146001.2 | NP_666113.2 | 2 - 3 | 254 | 65 | |
| GenBank mRNA | AK087586.1 | - | 2 - 3 | 252 | 65 | |
| AK166346.1 | - | 2 - 3 | 223 | 65 | ||
| BC017516.1 | - | 2 - 3 | 247 | 4504441 | 65 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC017516.1 transcripts.
Set Membership: |
|

Hip1
Genomic Map