| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1000011116.mm.1 |
NM_001285785.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011116.mm.1 |
NM_146011.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011116.mm.1 |
NM_146011.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011116.mm.1 |
TC1000001560.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1000011115.mm.1 |
NM_001285785.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011115.mm.1 |
NM_146011.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011115.mm.1 |
NM_146011.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1000011115.mm.1 |
TC1000001560.mm.1 |
MTA TC |