Assay Details
| Gene Symbol: Gabrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146017.3 | NP_666129.1 | 1 - 2 | 156 | 89 | |
| GenBank mRNA | AK028946.1 | - | 1 - 2 | 156 | 89 | |
| AK160577.1 | - | 1 - 2 | 160 | 89 | ||
| BC023693.1 | - | 1 - 2 | 143 | 5344479 | 89 | |
| BC025550.1 | - | 1 - 2 | 101 | 5320737 | 89 | |
| BC027245.1 | - | 1 - 2 | 119 | 3602400 | 89 | |
| BC031196.1 | - | 1 - 2 | 130 | 5059564 | 89 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK028946.1,AK160577.1,BC023693.1,BC025550.1,BC027245.1,BC031196.1,NM_146017.3 transcripts.
Set Membership: |
|

Gabrp
Genomic Map