Assay Details
| Gene Symbol: Gabrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146017.3 | NP_666129.1 | 3 - 4 | 368 | 98 | |
| GenBank mRNA | AK028946.1 | - | 3 - 4 | 368 | 98 | |
| AK160577.1 | - | 3 - 4 | 372 | 98 | ||
| BC023693.1 | - | 3 - 4 | 355 | 5344479 | 98 | |
| BC025550.1 | - | 3 - 4 | 313 | 5320737 | 98 | |
| BC027245.1 | - | 3 - 4 | 331 | 3602400 | 98 | |
| BC031196.1 | - | 3 - 4 | 342 | 5059564 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gabrp
Genomic Map