Assay Details
| Gene Symbol: Scrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146027.2 | NP_666139.1 | 3 - 4 | 462 | 53 | |
| GenBank mRNA | AK075869.1 | - | 3 - 4 | 426 | 53 | |
| BC010784.1 | - | 3 - 4 | 413 | 53 | ||
| BC021346.1 | - | 3 - 4 | 431 | 4163541 | 53 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966420 | BC021346.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966420 | BF464943.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966420 | NM_146027.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1966420 | AK075869.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246848 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 518457 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 101500 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165954 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162453 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178794 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966420 | BC021346.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966420 | BF464943.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966420 | NM_146027.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1966420 | AK075869.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048248 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048225 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048227 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28048223 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162453 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178794 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165954 | - | - |
| Whole Mouse Genome G4122A | A_51_P158598 | BC021346.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158598 | NM_146027.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158598 | AK075869.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P158598 | BC010784.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121218 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109027 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162453 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165954 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178794 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09161406 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09161414 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09161404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28048273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048277 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09161406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09464807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09496887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09496891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048242 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09161410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09496893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09496886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09007507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09584859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048251 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048268 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09584882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048279 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28048257 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10380725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380727 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380731 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380728 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10380723 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451822_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436012_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10380721 | - | - |
| Mouse Genome 430 2.0 Array | 1436012_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1436012_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1451822_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1451822_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217140_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 217140_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100014056.mm.1 | NM_146027.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014056.mm.1 | NM_146027.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100014056.mm.1 | TC1100001527.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436012_s_at | - | - |
| Mouse Expression Set 430 | 1436012_PM_s_at | - | - |
| Mouse Expression Set 430 | 1451822_PM_a_at | - | - |
| Mouse Expression Set 430 | 1451822_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451822_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436012_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451822_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436012_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436012_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451822_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436012_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451822_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 553824 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1980441 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1980441 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001980441 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2692341 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2692341 | - | - |
| mouse-6 v1.1 (Illumina) | scl40970.7.1_28-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1980441 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40970.7.1_28-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980441 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2692341 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2692341 | - | - |

Scrn2
Genomic Map