Assay Details
| Gene Symbol: Stac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146028.4 | NP_666140.1 | 2 - 3 | 881 | 66 | |
| GenBank mRNA | AJ608761.1 | - | 2 - 3 | 694 | 66 | |
| AK078132.1 | - | 2 - 3 | 882 | 66 | ||
| AK079357.1 | - | 2 - 3 | 739 | 66 | ||
| AK144433.1 | - | 2 - 3 | 737 | 66 | ||
| BC024864.1 | - | 2 - 3 | 787 | 5361431 | 66 | |
| BC029794.1 | - | 2 - 3 | 728 | 4949472 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stac2
Genomic Map