Assay Details
| Gene Symbol: Stac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146028.4 | NP_666140.1 | 3 - 4 | 974 | 61 | |
| GenBank mRNA | AJ608761.1 | - | 3 - 4 | 787 | 61 | |
| AK078132.1 | - | 3 - 4 | 975 | 61 | ||
| AK079357.1 | - | 3 - 4 | 832 | 61 | ||
| AK144433.1 | - | 3 - 4 | 830 | 61 | ||
| BC024864.1 | - | 3 - 4 | 880 | 5361431 | 61 | |
| BC029794.1 | - | 3 - 4 | 821 | 4949472 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stac2
Genomic Map