Assay Details
| Gene Symbol: Plekhh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146030.2 | NP_666142.1 | 5 - 6 | 1059 | 73 | |
| GenBank mRNA | AK154510.1 | - | 5 - 6 | 1075 | 73 | |
| AK164128.1 | - | 5 - 6 | 1076 | 73 | ||
| BC020025.1 | - | 5 - 6 | 1059 | 3490992 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plekhh3
Genomic Map